Learn More
Medchemexpress LLC HY-133680 5mg , β-Tocopherol CAS:16698-35-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1336805MG
Encompass_Preferred
HY-133680 5mg β-Tocopherol CAS: 16698-35-4 β-Tocopherol is an analogue of vitamin E, exhibits antioxidant properties. β-Tocopherol can inhibit tyrosinase activity and melanin synthesis. β-Tocopherol also can prevent the inhibition of cell growth and of PKC activity caused by d-alpha-tocopherol. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1766