Learn More
Medchemexpress LLC B022 | 1202764-53-1 | 99.4% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12050110MG
Encompass_Preferred
B022 is a potent, selective small-molecule inhibitor of NF-κB-inducing kinase (NIK) used for research into NF-κB signaling, inflammation, and cell death. The compound contains an alkyne group enabling click-chemistry conjugation and is supplied as a solid or as a DMSO solution for flexible experimental use.
  • Potent NIK inhibition (Ki = 4.2 nM; IC50 = 15.1 nM).
  • High purity suitable for biochemical and cellular assays.
  • Contains an alkyne handle for copper-catalyzed azide-alkyne cycloaddition.
  • Available as solid (multiple mg sizes) and as a 10 mM solution in DMSO.
  • Stable when stored under recommended conditions (powder and solution storage guidelines).
Catalog No. 50-002-31706