Learn More
MOLECULAR DEPOT LLC B12 Release Buffer - 50 mL
Supplier:  MOLECULAR DEPOT LLC B2010578
Encompass_Preferred
Vitamin B12 Release Buffer is an aqueous buffer for the fast and efficient release vitamin B12 from its serum/plasma transporter Unlike other release buffers this buffer is organic solvent-free and has been used in a wide array of biochemical and immunological applications related to vitamin B12 detection For in vitro research use only
Catalog No. 50-000-13501