Learn More
Medchemexpress LLC B7-1/CD80 Human HE 100ug
SDP
Supplier:  Medchemexpress LLC HYP7321100UG
Encompass_Preferred
B7-1/CD80 Protein Human (HEK293 Fc) is a polypeptide chain with the C-termimal human IgG1 Fc fragment produced in HEK293 cells CD80 is a costimulatory molecule known for its role in T-cell activation and also in regulating normal and malignant B cellsactivity
Catalog No. 50-001-82139