Learn More
Medchemexpress LLC B7-H2/ICOSLG Human 20ug | 20ug
SDP
Supplier:  Medchemexpress LLC HYP7807520UG
Encompass_Preferred
B7-H2/ICOSLG Protein a ligand for T-cell receptor ICOS critically costimulates T-cell proliferation and cytokine secretion It induces B-cell proliferation and supports plasma cell differentiation playing a key role in local tissue responses to inflammation B7-H2/ICOSLG also modulates the secondary immune response by co-stimulating memory T-cell function In molecular interactions it has been observed to interact with CTLA4 in vitro B7-H2/ICOSLG Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived B7-H2/ICOSLG protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87683