Learn More
ACROBiosystems ACROBiosystems Human B7-H3 (4Ig) / B7-H3b Protein, Fc Tag
Supplier:  ACROBiosystems B7BH5258100UG
Encompass_Preferred
Recombinant Protein;100UG;Human B7-H3 (4Ig), Fc Tag (B7B-H5258) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Thr 461 (Accession # Q5ZPR3-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.1 kDa. The protein migrates as 100-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-H3 (4Ig), Fc Tag (Cat. No. B7B-H5258) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 with a linear range of 0.3-5 ng/mL (QC tested).
Catalog No. 50-195-7138