Learn More
BPS Bioscience Inc B7-H4 Lentivirus 500 µl x 2
SDP
Supplier:  BPS Bioscience Inc 78727
Encompass
The B7-H4 Lentiviruses are replication incompetent, HIV-based, VSV-G pseudotyped lentiviral particles that are ready to transduce nearly all types of mammalian cells, including primary and non-dividing cells. The particles contain a human B7-H4 (NM_ 024626.4) driven by a CMV promoter and a puromycin selection marker
Catalog No. NC2980088