Learn More
Medchemexpress LLC B7-H4 protein (rat, HEK293, C-terminal His tag) | >95.0% | 100 UG

Supplier: Medchemexpress LLC HYP75485100UG
Recombinant rat B7-H4 protein expressed in HEK293 cells with a C-terminal 6xHis tag. The glycosylated protein is supplied lyophilized for use in immunology and cell-biology studies of T-cell regulation and immune checkpoint function.
- Expressed in HEK293 cells to preserve mammalian glycosylation.
- C-terminal 6xHis tag for purification and detection.
- Glycosylated molecular weight approximately 42-65 kDa.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Lyophilized from PBS (pH 7.4) for convenient storage and reconstitution.
- Available in multiple sizes including 100 μg.
- Store at -20°C; after reconstitution stable at 4°C for 1 week or at -20°C/-80°C for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.