Learn More
Medchemexpress LLC B7-H4 protein, rat (recombinant, HEK293, C-terminal His tag) | >95.0% | 10 UG
SDP
Supplier:  Medchemexpress LLC HYP7548510UG
Encompass_Preferred
Recombinant rat B7-H4 extracellular domain produced in HEK293 cells with a C-terminal His tag. Supplied lyophilized and purified for use in biochemical and immunological assays that require mammalian glycosylation and activity.
  • Produced in HEK293 cells with mammalian glycosylation.
  • C-terminal His tag for detection and purification.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Apparent molecular weight 42-65 kDa due to glycosylation.
  • Sequence length approximately 228 amino acids (extracellular domain).
  • Lyophilized from PBS, pH 7.4; aliquot and store at -20°C or -80°C after reconstitution.
  • Suitable for ELISA, western blot, and antibody binding studies.
Catalog No. 50-002-91402