Learn More
Medchemexpress LLC B7-H6 Human HEK293 5ug
SDP
Supplier:  Medchemexpress LLC HYP728395UG
Encompass_Preferred
B7-H6 a distinctive monomeric protein uniquely triggers NCR3-dependent natural killer (NK) cell activation specifically interacting with NCR3 while avoiding engagement with other NK cell-activating receptors (NCR1 NCR2 and KLRK1) This specificity underscores B7-H6 s unique role in initiating NK cell responses through the NCR3 pathway within the intricate network of NK cell activation mechanisms B7-H6 Protein Human (HEK293 Fc) is the recombinant human-derived B7-H6 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-81891