Learn More
Medchemexpress LLC B7-H6 protein, cynomolgus | >95.0% | 50 UG

Supplier: Medchemexpress LLC HYP7674350UG
Recombinant cynomolgus B7-H6 extracellular domain produced in HEK293 cells with a C-terminal 6xHis tag. Supplied lyophilized from 0.22 μm-filtered PBS (pH 7.4), this glycosylated protein corresponds to amino acids D25-D259 and has an apparent molecular weight of ~41-52 kDa. It is intended for research use in studies of natural killer cell activation and B7 family interactions.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Supplied lyophilized for long-term storage.
- Expressed in HEK293 mammalian cells for correct folding and glycosylation.
- Contains a C-terminal 6xHis tag for purification or detection.
- Amino acid range D25-D259 (extracellular domain).
- Suitable for assays studying NK cell activation and ligand interactions.
- Molecular weight approximately 41-52 kDa due to glycosylation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.