Learn More
Cayman Chemical B71CD80 ChImRIc MoncLNal 200ug
SDP
Supplier:  Cayman Chemical 37164200 UG
Encompass_Preferred
Immunogen: Recombinant human B7-1/CD80. The variable regions of the light and heavy chains were then cloned by incorporation into a cassette vector (N5LG1) containing human constant region genes and subsequently transfected into the Dg44 CHO cell line. • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) Human • Cross Reactivity: (+) CD80 • Applications: ELISA, FC, IHC, IP, WB, Functional assays
Catalog No. 50-291-9458