Learn More
OriGene B7H3 (CD276) Humanized Monoclonal Antibody, 100ug
SDP
Supplier:  OriGene TA355202
Encompass_Preferred
Applications- ELISA, FCRecommended Dilution- ELISA 1:5000-10000Reactivities- HumanHost- HumanizedIsotype- IgG1Clonality- MonoclonalFormulation- Lyophilized from sterile PBS, pH 7.4. Normally 5 % - 8 % trehalose is added as protectants before lyophilization.Preservative: 0.1% Procline 300
Catalog No. NC3313250