Learn More
ACROBiosystems Recombinant Protein;Human B7-H3 / CD276 Protein;HEK293;100UG;B73,B7-H3,CD276,B7 homolog 3,B7H3
Supplier: ACROBiosystems B73H52E2100UG
Recombinant Protein;Human B7-H3 / CD276 Protein;100UG;Human B7-H3, His Tag (B73-H52E2) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Pro 245 (Accession # Q5ZPR3-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.2 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.