Learn More
LGC GENOMICS LLC (Lucigen) BAC-Optimized Replicator v2.0 Electrocomp Cell, 24 Rxns (DUOs)
Supplier:  LGC GENOMICS LLC 602102
Encompass
BAC-Optimized Replicator™ v2.0 Electrocompetent Cells (> 1 10^10 cfu/g pKanR DNA). These cells are required for on-demand, copy number amplification of the pSMART BAC or pEZ™ BAC vectors in the CopyRight Kits, thereby increasing DNA yields. BAC-Optimized Replicator Cells are components of Lucigens CopyRight Cloning Kits.
Catalog No. NC9045708