Learn More
Bachem H-Arg(Pbf)-2-chlorotrityl resin (100-200 mesh, 0.50-0.90 mmol/g)
SDP
Supplier:  Bachem 4062605.0001
Encompass_Preferred
H-Arg(Pbf)-2-chlorotrityl resin (100-200 mesh, 0.50-0.90 mmol/g), 1g.
Catalog No. 50-194-7111