Learn More
ATCC Enterococcus faecalis; Quantitative DNA
Supplier:  ATCC 700802DQ
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-1120
 Restricted Item