Learn More
ATCC Bacillus cereus
Supplier:  ATCC 11778
Encompass
Bacillus cereus FDA strain PCI 213 [ATCC 9634, NCTC 10320, S.A. Waksman strain O] is a whole-genome sequenced bacterium with applications in media testing, quality control, and antibiotic assays.
Catalog No. 50-238-3405