Learn More
ATCC Human papilloma virus; type 16 clone; pHPV16 plasmid; in E. coli
Supplier:  ATCC 45113
Encompass
This is a clone of HPV16 that has been linearized at the unique BamHI site and cloned into the XhoI site of the pBluescript SK- vector.
Catalog No. 50-237-9612
 Restricted Item