Learn More
ATCC Bacillus spizizenii (Nakamura et al. 1999) Dunlap et al. 2020
Supplier:  ATCC 6633/DEL
Encompass
This item is used in sterility testing and assay of rifampin [rifampicin, rifamycin AMP], neomycin, monensin, kanamycin, lasalocid [X-537A], mitomycin [mitomycin C], vancomycin, novobiocin, penicillin, framycetin, tobramycin, spiramycin, gentamicins [gentamicin], amoxicillin, bekanamycin [kanamycin B; aminodeoxykanamycin], hygromycin B, streptomycin, erythromycin, dactinomycin [actinomycin; actinomycin D], streptonigrin, lactam antibiotics [beta-lactam antibiotics], capreomycin sulfate [capreomycin], and dihydrostreptomycin sulfate [dihydrostreptomycin]. Susceptibility disc testing of rifampin [rifampicin; rifamycin AMP] and novobiocin. Produces restriction endonuclease Bsu6633I and subtilin.
Catalog No. 50-189-127FP
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product