Learn More
Research Products International Corp Bacitracin, 5 Grams
SDP
Supplier:  Research Products International Corp B320005.0
Encompass_Preferred
Bacitracin is an antibiotic that has significant importance in the life science marketplace. It is derived from the bacterium Bacillus subtilis and belongs to the class of polypeptide antibiotics.
In the life science marketplace, bacitracin is primarily used for its antimicrobial properties. It is commonly employed in microbiology laboratories, research, and pharmaceutical industries. Bacitracin is effective against a wide range of Gram-positive bacteria, including Staphylococcus and Streptococcus species.
One of the key applications of bacitracin is in cell culture and tissue culture work. It is used as an additive in cell culture media to prevent bacterial contamination, ensuring the growth of uncontaminated cell lines. Bacitracin acts by inhibiting the synthesis of the bacterial cell wall, leading to the disruption of bacterial growth and preventing contamination.
Catalog No. 50-213-165