Learn More
Selleck Chemical LLC Bacitracin S4146-1g
SDP
Supplier:  Selleck Chemical LLC S41461G
Encompass_Preferred
Bacitracin is a mixture of related cyclic polypeptides produced by organisms of the licheniformis group of Bacillus subtilis var Tracy which disrupts both gram positive and gram negative bacteria by interfering with cell wall and peptidoglycan synthesis
Catalog No. 50-310-8700