Learn More
Medchemexpress LLC Bacitracin Zinc | 1405-89-6 | 98.76% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0278500MG
Encompass_Preferred
Bacitracin Zinc is a complex formed by the binding of Bacitracin with zinc ions. It is an orally active polypeptide antibiotic with bactericidal properties. This compound can cause DNA and deoxyribose damage, and it can also improve the gut microbiota of broiler and beef cattle.
  • Orally active polypeptide antibiotic
  • Bactericidal properties
  • Causes DNA and deoxyribose damage
  • Improves gut microbiota in broiler and beef cattle
Catalog No. 50-004-09002