Learn More
Biocare Medical, LLC Background Terminator - 25 ml
SDP
Supplier:  Biocare Medical, LLC BT967H
Encompass_Preferred
Background Terminator is the ultimate blocking reagent for many sources of nonspecific background staining in immunohistochemical techniques. Studies have shown it is superior to goat serum, dried milk, gelatin and BSA blocking reagents.
Catalog No. 50-823-89