Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC CAT Hedge Trimmer, 60V, 25 in, Brushless, 2-Speed, Backpack

Supplier:  MSC 64914534

Encompass

This 60V Backpack Hedge Trimmer delivers power and cutting capability for professional or homeowner use. Its backpack battery design makes it extremely light for extended use and precise shaping. Enjoy professional-level performance without the noise, complexity, or fumes of gas-powered hedge trimmers. It features a high-efficiency Brushless 2.0 motor, offering 40% more power and extended run time compared to brushed motor designs.

  • High-efficiency brushless 2.0 motor provides 40% more power and extended run time.
  • Ergonomic rotating rear handle allows comfortable cutting in any position.
  • Laser cut 25 inch dual edge blade ensures a clean cut.
  • Blade guard offers enhanced protection.
  • Achieve high performance without gas-related noise, maintenance, or fumes.
  • Backpack battery design reduces tool weight for easier trimming.

Catalog No. 50-283-7279


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.