Learn More
Grainger Backpack Vacuum, Corded, 120 cfm, HEPA Vacuum Filtration Type, 11.5 lb, 2 1/2 gal
Supplier:  Grainger 30RR53
Encompass_Preferred
Bagged, corded backpack vacuums trap dirt, dust, and allergens in replaceable bags that are discarded when full. They keep debris contained for disposal to help prevent recirculation of dust and allergens, making them a common choice for vacuuming in healthcare and educational settings. These backpack vacuums allow movement as far as the cord extends for cleaning underneath, around, and between furniture and equipment. Corded backpack vacuums plug into an electrical outlet and rest on your back as they remove dirt and dust from carpets and bare floors. They strap to you to be carried along for cleaning ceilings, walls, corners, and stairways. Their corded design eliminates the battery-recharging downtime associated with cordless units.
Catalog No. NC1892078