Learn More
Medchemexpress LLC Bacopaside II | 382146-66-9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN60165MG
Encompass_Preferred
Bacopaside II, extracted from the medicinal herb Bacopa monnieri, is an inhibitor of AQP1. It exhibits anti-angiogenesis activity and induces apoptosis in various cancer cells, demonstrating cytotoxicity against cancer cells.
  • Inhibits AQP1-mediated transport
  • Exhibits anti-angiogenesis activity
  • Induces apoptosis in cancer cells
  • Shows cytotoxicity against cancer cells
  • Inhibits proliferation of cancer cells (HT-29, SW480, SW620, HCT116) with IC50s ranging from 14.5 μM to 18.4 μM
  • Arrests cell cycle at G0/G1 phase (HT-29) or G2/M phase (SW480, SW620, HCT116)
Catalog No. 50-004-18479