Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bacopaside X | 94443-88-6 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN51405MG

Encompass_Preferred

Bacopaside X is found in *Bacopa monnieri* and demonstrates a binding affinity toward the D1 receptor. It has a molecular weight of 899.07 and a formula of C46H74O17. Bacopaside X appears as a white to off-white solid.

  • Purity: 99.64%
  • Target: D1 receptor
  • Structure classification: Terpenoids, Triterpenes
  • Initial source: Plants, Scrophulariaceae, *Bacopa monnieri* (Linn.) Wettst.

Catalog No. 50-004-20993


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.