Learn More
Medchemexpress LLC HY-N5064 5mg Medchemexpress, Bacoside A3 CAS:157408-08-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN50645MG
Encompass_Preferred
Medchemexpress, HY-N5064 5mg Bacoside A3 CAS:157408-08-7 Bacoside A3, a triterpenoid saponin, is one of the major active constituents in Bacopa monnieri. Bacoside A3 has neuroprotective activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2942