Learn More
Biotium Bacterial Viability Cell and Organelle Stains Nuclear Stains and DNA RNA Dyes Chemical Reagents 1/EA
Supplier:  Biotium 32020
Encompass_Preferred
Kit with Biotiums novel BactoView Dead 655/670 and BactoView Live Green Counterstain for two-color fluorescence staining of dead and live bacteria Provides better live/dead discrimination than traditional dyes for gram plus bacteria 100 uL each live cell stain (500X) and dead cell stain (1000X)
Catalog No. 50-005-01422
Only null left
Edge