Learn More
New England Biolabs, Inc. Bacteroides Heparinase II – 200 units
SDP
Catalog No. 50591172
Encompass
Bacteroides Heparinase II (also called Heparin Lyase II) is cloned from Bacteroides eggerthii. It is a low specificity enzyme that is active on both heparin and heparan sulfate. Bacteroides Heparinase II cleaves the glycosidic bond between N-sulfated and glucuronic or iduronic acid residues. The reaction yields oligosaccharide products containing unsaturated uronic acids which can be detected by UV spectroscopy at 232 nm. When used alone this enzyme rarely yields complete depolymerization of a polysaccharide chain, however disaccharide analysis is enhanced when used in combination with Heparinase I and III.
Catalog No. 50-591-172 Supplier New England Biolabs, Inc. Supplier No. P0736L
Bacteroides Heparinase II (also called Heparin Lyase II) is cloned from Bacteroides eggerthii. It is a low specificity enzyme that is active on both heparin and heparan sulfate. Bacteroides Heparinase II cleaves the glycosidic bond between N-sulfated and glucuronic or iduronic acid residues. The reaction yields oligosaccharide products containing unsaturated uronic acids which can be detected by UV spectroscopy at 232 nm. When used alone this enzyme rarely yields complete depolymerization of a polysaccharide chain, however disaccharide analysis is enhanced when used in combination with Heparinase I and III.