Learn More
ATCC Bacteroides thetaiotaomicron
Supplier:  ATCC 29148
Encompass
Bacteroides thetaiotaomicron strain VPI 5482 was isolated from human feces. This whole-genome sequenced bacterial type strain is propagated anaerobically.
Catalog No. 50-238-2889
 Restricted Item