Learn More
iQ Biosciences Bactrian Camel PBMCs, 100M cells/vial
Supplier:  iQ Biosciences IQBCMPB1100
Encompass_Preferred
High-quality, high-viability (> 80%) bactrian camel peripheral blood mononuclear cells (PBMCs) sourced responsibly from a vetted third party facility. Potential uses include, production of stable single-domain antibodies (nanobodies) for therapeutic and diagnostic applications, vaccine development, and autoimmune research. All orders come with an iQ Certificate of Analysis.
Catalog No. NC3235565