Learn More
Medchemexpress LLC BAD (103-127) (human) | 331762-68-6 | 98.80% | 3103.47 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP24685MG
Encompass_Preferred
BAD (103-127) (human) is a 25-mer Bad peptide derived from the BH3 domain of BAD. This peptide effectively antagonizes the function of Bcl-xL.
  • Derived from the BH3 domain of BAD.
  • Effectively antagonizes the function of Bcl-xL.
  • Exhibits almost 800-fold higher affinity for Bcl-XL compared to the 16-mer peptide.
Catalog No. 50-004-23897