Learn More
Cayman Chemical BaeocystIn-d4 1mg
SDP
Supplier:  Cayman Chemical 371251 MG
Encompass_Preferred
An internal standard for the quantification of baeocystin by GC- or LC-MS; an intermediate in the biosynthesis of psilocybin; intended for research and forensic applications
Catalog No. 50-291-4970