Learn More
Selleck Chemical LLC Siponimod (BAF312) 5mg 1230487-00-9
SDP
Supplier:  Selleck Chemical LLC S71795MG
Encompass_Preferred
BAF312 (Siponimod) is a next-generation S1P receptor agonist, selective for S1P1 and S1P5 receptors with EC50 of 0.39 nM and 0.98 nM, exhibits >1000-fold selectivity over S1P2, S1P3 and S1P4 receptors. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4495