Learn More
Selleck Chemical LLC Bafetinib S1369-25mg
SDP
Supplier:  Selleck Chemical LLC S136925MG
Encompass_Preferred
Bafetinib is a potent and selective dual Bcr-Abl/Lyn inhibitor with IC50 of 5 8 nM/19 nM in cell-free assays does not inhibit the phosphorylation of the T315I mutant and is less potent to PDGFR and c-Kit
Catalog No. 50-310-8156