Learn More
Alkali Scientific Baffled 1000mL Erlenmeyer Flask with Vented Cap and Flat Bottom, PETG, Sterile, 6/cs
SDP
Supplier:  Alkali Scientific EFB306
Encompass
Baffled interior bottoms provide a swirl diruption/break which improves the aeration of the culture. This feature can be beneficial to use when the cultures' oxygen requirements are high or viscous.
Catalog No. 50-233-5992