Learn More
Genscript Corporation BAFFR/TNFRSF13CBiotin His
Supplier:  Genscript Corporation Z04414500
Encompass
BAFF binds to three TNF receptor superfamily members B-cell maturation antigen (BCMA/TNFRSF17) transmembrane activator and calcium-modulator and cyclophilin ligand interactor (TACI/TNFRSF13B) and BAFF receptor (BAFF R/BR3/TNFRSF13C) These receptors are type III transmembrane proteins that lack a signal peptide Whereas TACI and BCMA bind BAFF and another TNF superfamily ligand APRIL (a proliferation-inducing ligand) BAFF R selectively binds BAFF
Catalog No. 50-000-59170