Learn More
Enzo Life Sciences Bafilomycin A1, (100µg), CAS Number: 88899-55-2
SDP
Supplier:  Enzo Life Sciences BMLCM1100100
Encompass_Preferred
ATPase inhibitor. Macrolide antibiotic that acts as a potent and selective inhibitor of vacuolar-type H+-ATPase (V-type) (IC50=004-0.4nmol/mg). Valuable tool for distinguishing among different types of ATPases. Inhibitor of endosomal acidification.¨Blocks lysosomal cholesterol trafficking in macrophages. Blocks pH regulation in brain cells. Formula: C35H58O9. MW: 622.8. CAS Number: 88899-55-2. Purity: ≥95% (HPLC, TLC). Appearance: White powder to colorless film. Solubility: Soluble in 100% ethanol, methanol or DMSO. Use/Stability: Stable for at least 1 year after receipt when stored at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C
Catalog No. 50-103-1771