Learn More
Cayman Chemical BafIlomycIn B1 100ug
SDP
Supplier:  Cayman Chemical 14005100 UG
Encompass_Preferred
A bacterial metabolite; inhibits the activity of rat liver lysosomal V-ATPase in a concentration-dependent manner; induces autophagosome accumulation in MCF-7 cells from 10-1,000 nM; inhibits chloroquine-induced apoptosis in primary CGNs at 1 nM; reduces viral genome copy numbers in the culture supernatant of Vero E6 cells infected with H1N1 influenza A
Catalog No. 50-295-3982