Learn More
Medchemexpress LLC Bafilomycin B1 | 88899-56-3 | 5 MG
Supplier: Medchemexpress LLC HYN67385MG
Bafilomycin B1 is a macrolide antibiotic isolated from the mycelium of Streptomyces gresuis. It is a structural analog of Bafilomycin A1 and functions as an inhibitor of V-ATPase. This compound has demonstrated neuroprotective properties against Chloroquine-induced cell death. Furthermore, Bafilomycin B1 significantly inhibits the accumulation of lipid droplets induced by oxidized low-density lipoprotein (LDL).
- Macrolide antibiotic: derived from Streptomyces gresuis mycelium.
- V-ATPase inhibitor: acts as an inhibitor of V-ATPase, a key enzyme involved in various cellular processes.
- Neuroprotective effects: provides neuroprotection against Chloroquine-induced neuronal death.
- Lipid accumulation inhibition: significantly reduces the accumulation of lipid droplets induced by oxidized LDL.
- Structural analog: shares structural similarities with Bafilomycin A1.