Learn More
MTC Bio Flip 'n Fold™ Sterile Sampling Bag, 4oz, 178mm x 76mm, 500/pk
Supplier:  MTC Bio B5760
Encompass_Preferred
With Flip 'n Fold™ Bags, samples can be easily collected by removing the tear strip above the wire closure and pulling on the two external grip tabs to open the mouth of the bag to the desired level. Using the grip tabs ensures sterility and eliminates the risk of contamination from finger contact. Then, introduce the sample into the bag, flip or fold the bag three times, bend the wires and your sample is secure inside the airtight and moisture sealed bag. Flip 'n Fold™ bags are made of virgin polyethylene for excellent clarity and strength. They meet USDA, EPA and FDA standards and are sterile, RNase-free, DNase-free and non-pyrogenic. The PVC tab that surrounds the stainless steel wire is extended beyond the wire ends to eliminate the potential for punctures, scratched hands or ripped gloves due to exposure to uncovered wire ends. Several sizes are supplied with a white printed area that eliminates tapes, labels, etc.
Catalog No. 50-226-8444