Learn More
Chemglass Life Sciences Chemglass Cell Culture Motion Bag, 20 L, Ultra Clear, 6-Port (CLS-1207-20)
SDP
Supplier:  Chemglass Life Sciences CLS120720
Encompass
Chemglass cell culture motion bag designed for rocker bioreactor systems. This single-use, 20 L bag is manufactured from ultra-clear robust film, supplied gamma irradiated, and engineered for extended mixing and gentle agitation of mammalian and plant cell cultures.
  • Capacity: 20 L.
  • Number of ports: 6.
  • Film material: ultra-clear robust film.
  • Sterilization: gamma irradiated.
  • Pack quantity: each; also available in packs of 10.
  • Typical applications: rocking bioreactors and gentle mixing of cell cultures.
Catalog No. NC3338373