Learn More
Chemglass Life Sciences Chemglass Bag, Cell Culture, Motion, 22 L (6-port) - for rocker bioreactor systems
SDP
Supplier:  Chemglass Life Sciences CLS120722M
Encompass
Single-use 22 L cell culture motion bag designed for rocker bioreactor systems. Made from ultra-clear film with low-density polyethylene product-contact surfaces; gamma irradiated and USP Class VI tested. Features six ports for tubing and fittings; available individually or in packs of 10.
  • 22 L capacity
  • Ultra-clear film construction
  • Low-density polyethylene product-contact surfaces
  • Six ports for tubing and fittings
  • Gamma irradiated for sterility
  • USP Class VI tested
  • Available individually or in packs of 10
Catalog No. 50-273-3327