Learn More
Chemglass Life Sciences Bag, Cell Culture, Motion, 50 L, Ultra Clear, CellGrow
SDP
Supplier:  Chemglass Life Sciences CLS120750
Encompass
Rocker cell culture bag designed for motion/rocker bioreactor systems that provides a 50 L working volume for mammalian and plant cell culture applications. The bag is made from ultra-clear multi-layer film (manufacturer lists RENOLIT 9101 / Flex 200 / CellGrow on product pages) and is supplied individually for single-use laboratory workflows.
  • 50 L capacity.
  • Designed for rocker-style cell culture systems.
  • Ultra clear film for visual monitoring of cultures.
  • Sold individually for single-use workflows.
  • Inner contact material is low density polyethylene.
Catalog No. NC3338356