Learn More
Electron Microscopy Sciences Drawstring Bead Bag for Thermal Microbead 5 L
SDP
Supplier:  Electron Microscopy Sciences 6860520
Encompass_Preferred
Nylon mesh bag with drawstring closure for storing, cleaning, using, and transporting thermal microbeads. The bag is designed to conveniently fit in ice buckets and store microbead in the freezer until ready for use. Place bag in an ice bucket on top of Cool Bricks for prolonged cooling.
Catalog No. 50-192-7579