Learn More
Selleck Chemical LLC BAI1 S8865-100MG
SDP
Supplier:  Selleck Chemical LLC S8865100MG
Encompass_Preferred
BAI1 is a direct allosteric inhibitor of BAX with a dissociation constant (Kd) of 15 0 4 M
Catalog No. 50-000-25388