Learn More
Apexbio Technology LLC Baicalein, 100mg Cas# 491-67-8
SDP
Supplier:  Apexbio Technology LLC N1858100
Encompass_Preferred
The flavonoid component of Nepalese and Sino-Japanese crude drugs.1,2 Baicalein,a major flavone of Scutellariae baicalensis,inhibits the 12-lipoxygenase (12-LOX) pathway of arachidonic acid metabolism,which inhibits cancer cell proliferation and induce Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8578