Learn More
Apexbio Technology LLC Baicalein 491-67-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC N18585.1
Encompass_Preferred
Baicalein is a natural flavonoid compound extracted from the medicinal herb Scutellaria baicalensis known to function as an inhibitor of lipoxygenase enzymes It selectively inhibits the 12-lipoxygenase (12-LOX) mediated metabolic pathway involved in arachidonic acid metabolism subsequently modulating inflammatory responses and cell proliferation signaling Baicalein demonstrates bioactivity as reflected by reported IC50 values typically ranging from 0 6 M to 2 5 M against purified human platelet 12-LOX enzyme It is utilized in biomedical research primarily to study inflammation pathways tumor cell proliferation processes and mechanisms underlying apoptosis induction
Catalog No. 50-000-63870